Anti-THRSP Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A64208-100UG A64208-50UG
77723-594EA 529.22 CAD
77723-594 77723-595
Anti-THRSP Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: THRSP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR