Anti-Scgb3a2 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A64182-100UG A64182-50UG
77723-545EA 529.22 CAD
77723-545 77723-546
Anti-Scgb3a2 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: Scgb3a2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR