Anti-Tmprss4 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A64114-100UG A64114-50UG
77723-414EA 529.22 CAD
77723-414 77723-415
Anti-Tmprss4 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: Tmprss4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR