Anti-ALMS1 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A64088-50UL A64088-100UL
77723-366EA 360.84 CAD
77723-366 77723-365
Anti-ALMS1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: ALMS1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR