Anti-ALMS1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A64085-100UL A64085-50UL
77723-359EA 529.22 CAD
77723-359 77723-360
Anti-ALMS1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IF:1:50-1:200

Type:
Antigen: ALMS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR