Anti-CMTM2 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A64075-100UG A64075-50UG
77723-340EA 529.22 CAD
77723-340 77723-341
Anti-CMTM2 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: CMTM2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR