Anti-CMTM2 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A64074-50UG A64074-100UG
77723-339EA 360.84 CAD
77723-339 77723-338
Anti-CMTM2 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: CMTM2
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR