Anti-EXPH5 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A64016-50UG A64016-100UG
77723-233EA 360.84 CAD
77723-233 77723-232
Anti-EXPH5 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: EXPH5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR