Anti-FBXO15 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63992-100UG A63992-50UG
77723-187EA 529.22 CAD
77723-187 77723-188
Anti-FBXO15 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: FBXO15
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR