Anti-FSIP1 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63956-50UG A63956-100UG
77723-121EA 360.84 CAD
77723-121 77723-120
Anti-FSIP1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: FSIP1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR