Anti-C8orf31 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63948-100UG A63948-50UG
77723-105EA 529.22 CAD
77723-105 77723-106
Anti-C8orf31 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: C8orf31
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR