Anti-RAB42 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG BIOSCIENCE LLC
A63905-100UG A63905-50UG
77723-023EA 529.22 CAD
77723-023 77723-024
Anti-RAB42 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: RAB42
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR