Anti-SLC35F6 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63888-100UG A63888-50UG
77722-991EA 529.22 CAD
77722-991 77722-992
Anti-SLC35F6 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: SLC35F6
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR