Anti-LYG1 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63879-100UG A63879-50UG
77722-975EA 529.22 CAD
77722-975 77722-976
Anti-LYG1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: LYG1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR