Anti-PPP1R16B Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63863-100UG A63863-50UG
77722-944EA 529.22 CAD
77722-944 77722-945
Anti-PPP1R16B Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: PPP1R16B
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR