Anti-FYTTD1 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63836-50UG A63836-100UG
77722-894EA 360.84 CAD
77722-894 77722-893
Anti-FYTTD1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: FYTTD1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR