Anti-FYTTD1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A63833-100UG A63833-50UG
77690-312EA 529.22 CAD
77690-312 77722-888
Anti-FYTTD1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:2000

Type:
Antigen: FYTTD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR