Anti-SLC4A9 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63832-50UG A63832-100UG
77722-887EA 360.84 CAD
77722-887 77722-886
Anti-SLC4A9 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: SLC4A9
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR