Anti-TSGA13 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63823-100UG A63823-50UG
77722-869EA 529.22 CAD
77722-869 77722-870
Anti-TSGA13 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: TSGA13
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR