Anti-C9orf72 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63770-50UG A63770-100UG
77722-768EA 360.84 CAD
77722-768 77722-767
Anti-C9orf72 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: C9orf72
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR