Anti-SIGLEC10 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63766-100UG A63766-50UG
77722-759EA 529.22 CAD
77722-759 77722-760
Anti-SIGLEC10 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: SIGLEC10
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR