Anti-MAEL Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63754-50UG A63754-100UG
77722-737EA 360.84 CAD
77722-737 77722-736
Anti-MAEL Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: MAEL
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR