Anti-SPG11 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63750-50UG A63750-100UG
77722-729EA 360.84 CAD
77722-729 77722-728
Anti-SPG11 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: SPG11
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR