Anti-CDK5RAP3 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63746-50UL A63746-100UL
77722-721EA 360.84 CAD
77722-721 77722-720
Anti-CDK5RAP3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: CDK5RAP3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR