Anti-SPRR2A Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63696-50UG A63696-100UG
77722-626EA 360.84 CAD
77722-626 77722-625
Anti-SPRR2A Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: SPRR2A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR