Anti-VTI1A Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63655-100UG A63655-50UG
77722-544EA 529.22 CAD
77722-544 77722-545
Anti-VTI1A Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: VTI1A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR