Anti-YIPF5 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A63629-50UG A63629-100UG
77722-494EA 360.84 CAD
77722-494 77722-493
Anti-YIPF5 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: YIPF5
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR