Anti-MYDGF Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A63627-100UL A63627-50UL
77690-297EA 529.22 CAD
77690-297 77722-491
Anti-MYDGF Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:200-1:1000, IHC:1:20-1:200

Type:
Antigen: MYDGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR