Anti-GHSR Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A63610-100UG A63610-50UG
77722-457EA 529.22 CAD
77722-457 77722-458
Anti-GHSR Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: GHSR
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR