Anti-NP Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63575-100UG A63575-50UG
77722-391EA 529.22 CAD
77722-391 77722-392
Anti-NP Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: NP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Reston ebolavirus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR