Anti-CLEC4D Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A63555-100UG A63555-50UG
77722-354EA 529.22 CAD
77722-354 77722-355
Anti-CLEC4D Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: CLEC4D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR