Anti-SMAD1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A63547-50UL A63547-100UL
77722-342EA 360.84 CAD
77722-342 77722-341
Anti-SMAD1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:50-1:200

Type:
Antigen: SMAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR