Anti-MUC13 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48534-100UL A48534-50UL
77714-844EA 529.22 CAD
77714-844 77714-845
Anti-MUC13 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:15-1:50

Type:
Antigen: MUC13
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR