Anti-FPGS Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48517-100UG A48517-50UG
77689-272EA 529.22 CAD
77689-272 77714-816
Anti-FPGS Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000

Type:
Antigen: FPGS
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR