Anti-ATF1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48471-50UL A48471-100UL
77714-733EA 360.84 CAD
77714-733 77714-732
Anti-ATF1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: ATF1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR