Anti-LRP6 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48444-50UL A48444-100UL
77714-690EA 360.84 CAD
77714-690 77714-689
Anti-LRP6 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:40-1:200

Type:
Antigen: LRP6
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR