Anti-COG6 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A48437-100UG A48437-50UG
77714-678EA 529.22 CAD
77714-678 77714-679
Anti-COG6 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: COG6
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR