Anti-COG6 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48434-50UG A48434-100UG
77714-673EA 360.84 CAD
77714-673 77714-672
Anti-COG6 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:500-1:1000

Type:
Antigen: COG6
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR