Anti-FOXN1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48408-100UL A48408-50UL
77714-624EA 529.22 CAD
77714-624 77714-625
Anti-FOXN1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:25-1:100

Type:
Antigen: FOXN1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR