Anti-TERT Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48373-100UG A48373-50UG
77705-366EA 400.93 CAD
77705-366 77714-564
Anti-TERT Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, IHC:1:50-1:300, ELISA:1:5000-1:20000

Type:
Antigen: TERT
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR