Anti-OCC1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48371-100UL A48371-50UL
77714-560EA 529.22 CAD
77714-560 77714-561
Anti-OCC1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: OCC1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR