Anti-XKRX Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48364-50UL A48364-100UL
77714-548EA 360.84 CAD
77714-548 77714-547
Anti-XKRX Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100

Type:
Antigen: XKRX
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR