Anti-PKD1 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A48360-50UG A48360-100UG
77714-541EA 360.84 CAD
77714-541 77714-540
Anti-PKD1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: PKD1
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR