Anti-FUS Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48297-100UL A48297-50UL
77714-424EA 529.22 CAD
77714-424 77714-425
Anti-FUS Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:30-1:150

Type:
Antigen: FUS
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR