Anti-ITIH3 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A48246-100UG A48246-50UG
77714-331EA 529.22 CAD
77714-331 77714-332
Anti-ITIH3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: ITIH3
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR