Anti-CLDN11 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48226-100UL A48226-50UL
77714-296EA 529.22 CAD
77714-296 77714-297
Anti-CLDN11 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: CLDN11
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR