Anti-PDE1A Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A48196-50UL A48196-100UL
77714-244EA 360.84 CAD
77714-244 77714-243
Anti-PDE1A Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: PDE1A
Clonality: Polyclonal
Clone:
Conjucation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR