Anti-ATG4A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48190-100UG A48190-50UG
77705-365EA 400.93 CAD
77705-365 77714-233
Anti-ATG4A Rabbit Polyclonal Antibody
Antibodies
IHC:1:50-1:200, ELISA:1:10000

Type:
Antigen: ATG4A
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR