Anti-ERI1 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A48086-100UG A48086-50UG
77714-050EA 529.22 CAD
77714-050 77714-051
Anti-ERI1 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: ERI1
Clonality: Polyclonal
Clone:
Conjucation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR