Anti-ACO1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48041-100UL A48041-50UL
77713-962EA 529.22 CAD
77713-962 77713-963
Anti-ACO1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, WB:1:1000-1:5000

Type:
Antigen: ACO1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR